SHOWA CharGuard Heat-Resistant Gloves

High heat resistance up to 500°F (260°C) with superior dexterity
Ideal for use with autoclaves, food service, inspection, and more! Thermal-protective gloves feature a non-woven liner for excellent flexibility and fit. Designed to withstand heat exposure and maintain superior tactile dexterity, even after multiple washings. Textured gloves provide an excellent grip. The black color doesn't show dirt and grime extending its wearable life.
2,600.00 - ₹5,000.00INR / Pair of 1

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
SHOWA 8814-7 Heat Resistant Glove, 14"L, Size 7 WW-86483-85 Mfr # 8814-7
COMPARE
14 7
₹4,000.00INR / Pair of 1
SHOWA 8814-8 Heat Resistant Glove, 14"L, Size 8 WW-86483-86 Mfr # 8814-8
COMPARE
14 8
₹2,600.00INR / Pair of 1
SHOWA 8814-9 Heat Resistant Glove, 14"L, Size 9 WW-86483-87 Mfr # 8814-9
COMPARE
14 9
₹5,000.00INR / Pair of 1
SHOWA 8814-10 Heat Resistant Glove, 14"L, Size 10 WW-86483-88 Mfr # 8814-10
COMPARE
14 10
₹5,000.00INR / Pair of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat