SHOWA Butyl II Gloves
Perfect for chemical handling, spill control, and hazmat applications
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
7,900.00 - ₹10,800.00INR / Pair of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Grip Size Thickness (mil)
Availability Pricing
SHOWA 874-08 Gloves, smooth, medium WW-86357-01 Mfr # 874-08
COMPARE
Smooth Medium 14
₹7,900.00INR / Pair of 1
SHOWA 874-09 Gloves, smooth, 14 mil, large WW-86357-02 Mfr # 874-09
COMPARE
Smooth Large 14
₹7,900.00INR / Pair of 1
SHOWA 874-10 Gloves, smooth, extra-large WW-86357-04 Mfr # 874-10
COMPARE
Smooth X-Large 14
₹7,900.00INR / Pair of 1
SHOWA 874R-8 Gloves, textured, medium WW-86357-11 Mfr # 874R-8
COMPARE
Textured Medium 14
₹8,300.00INR / Pair of 1
SHOWA 874R-9 Gloves, textured, large WW-86357-12 Mfr # 874R-9
COMPARE
Textured Large 14
₹8,300.00INR / Pair of 2
SHOWA 874R-10 Gloves, textured, extra-large WW-86357-13 Mfr # 874R-10
COMPARE
Textured X-Large 14
₹8,300.00INR / Pair of 2
SHOWA 878-08 Gloves, heavy-weight, medium WW-86357-20 Mfr # 878-08
COMPARE
Smooth Medium 30
₹10,800.00INR / Pair of 2
SHOWA 878-09 Gloves, Heavy-Weight, 28 mL Thickness, Large WW-86357-21 Mfr # 878-09
COMPARE
Smooth Large 28
₹10,800.00INR / Pair of 2
SHOWA 878-10 Gloves, heavy-weight, extra-large WW-86357-22 Mfr # 878-10
COMPARE
Smooth X-Large 30
₹10,800.00INR / Pair of 2
Showing 1- 9 of 9