Benchmark Scientific BeadBug™ 6 Microtube Homogenizer
The perfect fit for the needs of medium throughput laboratories
  • Sealed homogenization prevents cross-contamination
  • Process up to six 2-mL tubes simultaneously
  • Most samples can be processed within 45 seconds
  • Handles a variety of samples from cells to organ tissue to bone
The BeadBug 6 homogenizer uses a three-dimensional shaking motion. This motion results in high velocity impacts between the sample and the beads. This method is efficient for releasing cellular contents, such as nucleic acids and proteins. The digital display is simple to understand; program options include set speed, time, number of cycles, and pause between cycles. The last programmed parameters are stored in memory so you can easily do repeat runs.
523,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Max Speed (rpm) Power (VAC)
Availability Pricing
Benchmark Scientific Beadbug 6 Microtube Homogenizer, 115 VAC WW-36271-04 Mfr # D1036
COMPARE
2 4350 115
₹523,000.00INR / Each
Benchmark Scientific Beadbug 6 Microtube Homogenizer, 230 VAC WW-36271-05 Mfr # D1036-E
COMPARE
2 4350 230
₹523,000.00INR / Each