Bel-Art Robotic Sterile Filtered Pipette Tips
Ideal for use with Hamilton® and Tecan® robotic systems
  • DNase, RNase, and pyrogen free
  • Manufactured in an ISO Class 8 cleanroom
  • Filtered to prevent contamination of pipette arm
These sterile, disposable, filtered pipette tips can be use with automated liquid handling equipment as well as with Hamilton® and Tecan® robotic systems. The filtered tips protect pipette arm from cross-contamination.
Available in conductive and non-conductive tips. The conductive tips detect the liquid level when pipetting conductive liquids to ensure repeatable volume accuracy.
73,200.00 - ₹134,000.00INR / Pkg of 24
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compatible with Max Volume (µL) Tip Style
Availability Pricing
Bel-Art Robotic Sterile Filtered Pipette Tips, Hamilton Type, 50uL, Non-Conductive; 96 Tips/Rack x 24/PK WW-07898-05 Mfr # 378601005
COMPARE
Hamilton robotic systems 50 Non-conductive
₹113,000.00INR / Pkg of 24
Bel-Art Robotic Sterile Filtered Pipette Tips, Hamilton Type, 1000uL, Non-Conductive; 96 Tips/Rack x 16/PK WW-07898-06 Mfr # 378601100
COMPARE
Hamilton robotic systems 50 Non-conductive
₹83,800.00INR / Pkg of 16
Bel-Art Robotic Sterile Filtered Pipette Tips, Tecan Type, 50uL, Non-Conductive; 96 Tips/Rack x 24/PK WW-07898-07 Mfr # 378602005
COMPARE
Tecan robotic systems 1000 Non-conductive
₹100,000.00INR / Pkg of 24
Bel-Art Robotic Sterile Filtered Pipette Tips, Tecan Type, 1000uL, Non-Conductive; 96 Tips/Rack x 16/PK WW-07898-08 Mfr # 378602100
COMPARE
Tecan robotic systems 1000 Non-conductive
₹73,200.00INR / Pkg of 16
Bel-Art Robotic Sterile Filtered Pipette Tips, Hamilton Type, 50uL, Conductive; 96 Tips/Rack x 24/PK WW-07898-09 Mfr # 378603005
COMPARE
Hamilton robotic systems 50 Conductive
₹134,000.00INR / Pkg of 24
Bel-Art Robotic Sterile Filtered Pipette Tips, Hamilton Type, 1000uL, Conductive; 96 Tips/Rack x 16/PK WW-07898-10 Mfr # 378603100
COMPARE
Hamilton robotic systems 1000 Conductive
₹97,400.00INR / Pkg of 16
Bel-Art Robotic Sterile Filtered Pipette Tips, Tecan Type, 50uL, Conductive; 96 Tips/Rack x 24/PK WW-07898-11 Mfr # 378604005
COMPARE
Tecan robotic systems 50 Conductive
₹129,000.00INR / Pkg of 24
Bel-Art Robotic Sterile Filtered Pipette Tips, Tecan Type, 1000uL, Conductive; 96 Tips/Rack x 16/PK WW-07898-12 Mfr # 378604100
COMPARE
Tecan robotic systems 1000 Conductive
₹96,500.00INR / Pkg of 16
Showing 1- 8 of 8