This product is now discontinued, View Related Products

Avanti Basic-Use Refrigerators and Freezers

Economical models ideal for noncritical storage and general-purpose use
  • Perfect for general-purpose use from the lab to the breakroom
These high-quality units provide an economical alternative to lab refrigerators and freezers.
Call For Price

19 variations of this product are available.

See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Refrigerator, Basic-use 1.8 cu ft. with mini ice tray WW-44199-00 Mfr # GMR02WH
COMPARE
115
Call For Price
Avanti Undercounter Refrigerator, 1.7 cu ft, 110 VAC WW-44199-01 Mfr # RM17T0WH
COMPARE
110 1.7
Call For Price
Refrigerator, Basic-use 4.3 cu ft. with mini ice tray WW-44199-02 Mfr # GMR04WH
COMPARE
208 / 230
Call For Price
GMR06WH Refrigerator, Basic-use 6.0 cu ft. with mini ice tray WW-44199-04 Mfr # GMR06WH
COMPARE
115
Call For Price
FUM5SNWH Freezer, Basic-use undercounter, 5.0 cu ft. WW-44199-06 Mfr # FUM5SNWH
COMPARE
115
Call For Price
Freezer, Upright, Basic-use 13.7 cu ft. WW-44199-08 Mfr # AFFU1466
COMPARE
115
Call For Price
Freezer, Upright, Basic-use 16.7 cu ft. WW-44199-10 Mfr # AFFU1766
COMPARE
220
Call For Price
Freezer, Upright, Basic-use 16.7ft3 WW-44199-11 Mfr # FFU17F5
COMPARE
208 / 230
Call For Price
Freezer, Upright, Basic-use 20.0 cu ft. WW-44199-12 Mfr # AFFU2066
COMPARE
115
Call For Price
Freezer, Upright, Basic-use 20.6ft3 WW-44199-13 Mfr # FFFU21M1QW
COMPARE
220
Call For Price
Refrigerator/Freezer, Basic-use 11.7 cuft WW-44199-14 Mfr # GTS12BBRWH
COMPARE
208 / 230
Call For Price
Refrigerator/Freezer, Basic-Use; 14.6 cu ft. WW-44199-16 Mfr # GTE15CTHRWW
COMPARE
115 10.6
Call For Price
FFU14F7H Upright Freezer, 13.7 cu ft, 3 shelves, 1 basket, 120 VAC WW-44199-17 Mfr # FFU14F5HW
COMPARE
220
Call For Price
Refrigerator/Freezer, Basic-use 17.9 cu ft. Sku4419918 WW-44199-18 Mfr # GTS18KBWH
COMPARE
208 / 230
Call For Price
Refrigerator/Freezer, Basic-use 21.7 cu ft. WW-44199-20 Mfr # GTS22KBWH
COMPARE
Call For Price
Refrigerator/Freezer, Basic-use 17.9 cu ft. Sku4419921 WW-44199-21 Mfr # GTH18GBDWW
COMPARE
Call For Price
General Electric GTH18GBWH Refrigerator/Freezer, Basic-use 18.1 cu ft., White WW-44199-22 Mfr # GTS18GTHWH
COMPARE
115 13.89
Call For Price
General Electric GTH16BBRWH Refrigerator/Freezer, Basic-use 21 cu ft., White WW-44199-23 Mfr # GTH21GBEWW
COMPARE
Call For Price
Avanti RM4436SS 4.4 cu ft. Undercounter Refrigerator, Black with Stainless Steel Door WW-44199-40 Mfr # RM4436SS
COMPARE
110 4.4
Call For Price
Showing 1- 19 of 19

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat