This product is now discontinued, View Related Products
Baritainer Jerry Can, Natural, HDPE/Quoral
Environmentally friendly alternative to glass, metal, or fluorinated containers!
Manufactured of Quoral® and HDPE, these rugged carboys can be used to safely transport industrial and laboratory chemicals, solvents, adhesives, and cleaning solvents. Quoral is an additive that is added to high density polyethylene (HDPE) that utilizes laminate technology. The technology consists of the production of multiple overlapping layers of barrier material in the polyethylene
matrix. Temperature range is -148 to 248° F (-100 to 120°C).
3,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Baritainer Jerry Can, Natural, HDPE/Quoral 5 L WW-06069-01 Mfr # 405404-0005
COMPARE
5 DIN 50 High Density Polyethyene (HDPE)
Call For Price
Baritainer Jerry Can, Natural, HDPE/Quoral 10 L WW-06069-03 Mfr # 405594-0002
COMPARE
10 DIN 50 High Density Polyethyene (HDPE)
₹3,700.00INR / Each
Baritainer Jerry Can, Natural, HDPE/Quoral 20 L WW-06069-05 Mfr # 405594-2070
COMPARE
20 70 mm High Density Polyethyene (HDPE)