This product is now discontinued, View Related Products
Bacharach Monoxor Plus Carbon Monoxide Analyzers
Real-time display and trending graph provide data and analysis onsite
  • B-Smart® pre-calibrated sensors enable field replacement without downtime
  • Measure ambient and flue gas temperatures using any type K thermocouple with miniconnector
  • Internal diaphragm pump and detachable probe with hose allow monitoring of flues, furnaces, appliances and boilers
  • Automatic shut-off function powers down the analyzer after a user-selectable period of inactivity, unless carbon monoxide levels are above 50 ppm
  • Audible alarm sounds when CO levels exceed user defined limits
  • Memory holds up to 10 test records
The advanced easy-to-use Monoxor Plus carbon monoxide analyzer is perfect for contractors, safety technicians, and weatherization specialists. The B-Smart sensors provide fast and accurate readings, while the graph function shows trends over a selectable sampling period, making on-the-job analysis quick and painless. Use the differential temperature function for combustion applications. Once calculated, the differential temperature can be saved, displayed and printed for your records.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Type Gases Measured Description
Availability Pricing
Bacharach Monoxor Plus Carbon Monoxide Analyzer WW-10110-37 Mfr # H0019-8117
COMPARE
4 x AA batteries CO Carbon Monoxide Analyzer
Call For Price
Bacharach Monoxor Plus Carbon Monoxide Analyzer Kit WW-10110-55 Mfr # 0019-8118
COMPARE
4 x AA batteries CO Carbon Monoxide Analyzer Kit