This product is now discontinued, View Related Products
BabyBio™ Prepacked Immobilized Metal Ion Affinity Chromatography (IMAC) Columns
Prepacked columns provide a quick and convenient purification of polyhistidine-tagged proteins
  • High binding capacity and purity
  • Columns precharged with different metals or uncharged to accept ion of your choice
Quickly and easily purify his-tagged proteins using BabyBio IMAC columns. Columns are packed with WorkBeads™ agarose media with chelating ligands based on tris(2-aminoethyl)amine (TREN), iminodiacetic acid (IDA), or nitrilotriacetic acid (NTA). Select from uncharged TREN, or charged or uncharged IDA and NTA. Charged media contains IDA or NDA with Ni, Co, Cu, or Zn ions, which are immobilized on the media via the chelating ligand. Each column has a dynamic binding capacity of 50 mg/mL of medium. Connect columns in series for larger sample volumes.
Order BabyBio screening kits to conveniently repeat the same purification procedure with different metals to reduce the binding of host cell proteins or to optimize purity and yield. Each kit comes with four columns prepacked with WorkBeads IDA or WorkBeads NTA media precharged with Ni2+, Co2+, Cu2+, or Zn2+ ions.
Columns feature biocompatible polypropylene body and polyethylene top and bottom filters. Stable ove pH 7 to 9 range short term, 2 to 12 cleaning (stripped column). Each column comes with a plug in the inlet and a snap-off end at the outlet. Easily connect to a syringe, pump, or chromatography system using fingertight fittings (coned 10-32) for 1/16" OD tubing.
11,200.00 - ₹282,000.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 1 mL WW-11947-96 Mfr # KN45655001
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 1 mL
₹11,200.00INR / Each
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 2 x 1 mL WW-11947-97 Mfr # KN45655002
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 2 x 1 mL
₹20,000.00INR / Pkg of 2
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 x 1 mL WW-11947-98 Mfr # KN45655003
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 x 1 mL
₹38,800.00INR / Pkg of 5
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 10 x 1 mL WW-11947-99 Mfr # KN45655004
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 10 x 1 mL
₹70,200.00INR / Pkg of 10
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 mL WW-11948-00 Mfr # KN45655005
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 mL
₹34,300.00INR / Each
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 2 x 5 mL WW-11948-01 Mfr # KN45655006
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 2 x 5 mL
₹61,100.00INR / Pkg of 2
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 x 5 mL WW-11948-02 Mfr # KN45655007
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 5 x 5 mL
₹129,000.00INR / Pkg of 5
Bio-Works BabyBio Ni-IDA Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 10 x 5 mL WW-11948-03 Mfr # KN45655008
COMPARE
Pre-Packed Chromatography Column, Ni²⁺ Iminodiacetic Acid; 10 x 5 mL
₹230,000.00INR / Pkg of 10
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 1 mL WW-11948-04 Mfr # KN45655011
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 1 mL
₹11,200.00INR / Each
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 2 x 1 mL WW-11948-05 Mfr # KN45655012
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 2 x 1 mL
₹20,000.00INR / Pkg of 2
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 x 1 mL WW-11948-06 Mfr # KN45655013
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 x 1 mL
₹38,800.00INR / Pkg of 5
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 10 x 1 mL WW-11948-07 Mfr # KN45655014
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 10 x 1 mL
₹70,000.00INR / Pkg of 10
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 mL WW-11948-08 Mfr # KN45655015
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 mL
₹34,300.00INR / Each
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 2 x 5 mL WW-11948-09 Mfr # KN45655016
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 2 x 5 mL
₹61,100.00INR / Pkg of 2
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 x 5 mL WW-11948-10 Mfr # KN45655017
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 5 x 5 mL
₹129,000.00INR / Pkg of 5
Bio-Works BabyBio IDA Pre-Packed Chromatography Column, Iminodiacetic Acid; 10 x 5 mL WW-11948-11 Mfr # KN45655018
COMPARE
Pre-Packed Chromatography Column, Iminodiacetic Acid; 10 x 5 mL
₹231,000.00INR / Pkg of 10
Bio-Works BabyBio Cu-IDA Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 1 mL WW-11948-12 Mfr # KN45655021
COMPARE
Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 1 mL
₹11,200.00INR / Each
Bio-Works BabyBio Cu-IDA Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 2 x 1 mL WW-11948-13 Mfr # KN45655022
COMPARE
Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 2 x 1 mL
₹20,000.00INR / Pkg of 2
Bio-Works BabyBio Cu-IDA Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 5 x 1 mL WW-11948-14 Mfr # KN45655023
COMPARE
Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 5 x 1 mL
₹39,000.00INR / Pkg of 5
Bio-Works BabyBio Cu-IDA Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 10 x 1 mL WW-11948-15 Mfr # KN45655024
COMPARE
Pre-Packed Chromatography Column, Cu²⁺ Iminodiacetic Acid; 10 x 1 mL
₹70,400.00INR / Pkg of 10
Showing 1 - 20 of 50