This product is now discontinued, View Related Products
Bürkle Stainless Steel Mini ViscoSamplers
Sample highly viscous samples with ease
  • Ideal for pharmaceutical and cosmetics applications with the easy to clean design
  • Suitable for steam sterilization up to 250°F (121°C)
Samplers are suitable for viscous samples such as pastes, mud, creams, and oils. There are three removable tip sizes to choose from, with 2 mm to 6 mm holes, depending on the viscosity of the samples. The sample is drawn into the pump rod via suction and transported directly to the sampling container. This allows for clean and easy sampling, and provides easy clean up.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (in) Volume (mL) Description
Availability Pricing
Burkle 5341-1060 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 30 mL WW-06041-92 Mfr # 5341-1060
COMPARE
23.6 30 Burkle, Inc. 5341-1060 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 30 mL
Call For Price
Burkle 5341-1100 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 50 mL WW-06041-93 Mfr # 5341-1100
COMPARE
39.3 50 Burkle, Inc. 5341-1100 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 50 mL
Call For Price
Burkle 5341-1150 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 75 mL WW-06041-94 Mfr # 5341-1150
COMPARE
59 75 Burkle, Inc. 5341-1150 Small viscous substance sampling rod, manual draw, SS, 2 mm suction tip, 60 cm; 75 mL