This product is now discontinued, View Related Products
Bürkle Stainless Steel Large-Diameter Bulk Good Sampling Lances
Useful for large-sized goods up to 1 cm in diameter
  • Ideal for hazelnuts, blends of tea, grain, or coarse-grained bulk goods
  • Corresponds to ISTA standard for seed lengths of 10 mm (wheat, oats) and 5 mm (flax, vetches)
  • Suitable for steam sterilization up to 250°F (121°C)
  • No-hassle maintenance-free sampler
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Material
Availability Pricing
Burkle 5382-0040 Tube sampler for bulk goods, SS, 40 cm x 40 mm, meets ISTA standards WW-06042-96 Mfr # 5382-0040
COMPARE
1.6 15.8 316L Stainless Steel and PP
Call For Price
Burkle 5382-0080 Tube sampler for bulk goods, SS, 80 cm x 40 mm, meets ISTA standards WW-06042-97 Mfr # 5382-0080
COMPARE
1.6 31.5 316L Stainless Steel and PP