This product is now discontinued, View Related Products
Bürkle Silo Drill Sampler for Bulk Goods
Efficiently take samples from different depths of your bulk goods silo
  • Extension arm available for hard-to-reach depths
  • Suitable for steam sterilization up to 250°F (121°C)
  • No-hassle maintenance-free samplers
The silo drill sampler is suitable for bulk goods that have been transferred over long distances in which separation may occur—it allows for representative samples to be taken from all layers, eliminating incorrect results obtained from sample surfacing only. The collection cylinder remains closed as the sampler is screwed into the sample and opens easily with the turn of the handle at the desired sample depth. For easy boring, this unit features a sturdy drill tip with drilling blades. The standard length of the sampler is 150 cm, with optional extension arm allowing for a maximum length of 350 cm.
3 PRODUCT OPTIONS
Compareitem specifications
Material Volume (mL) Description
Availability Pricing
Burkle 5301-0500 Silo Drill Sampler Extension, Stainless Steel; 100 cm WW-06040-56 Mfr # 5301-0500
COMPARE
304 Stainless Steel Burkle, Inc. 5301-0500 Silo drill sampler extension, SS, 100 cm
Call For Price
Burkle 5301-1000 Silo Drill Sampler, Aluminum; 400 mL WW-06040-57 Mfr # 5301-1000
COMPARE
Aluminium 400 Burkle, Inc. 5301-1000 Silo drill sampler, manual operation, aluminum, 150 cm long, 90 mm screw, 40 mm chamber; 400 mL
Call For Price
Burkle 5301-2000 Silo Drill Sampler, Stainless Steel; 400 mL WW-06040-58 Mfr # 5301-2000
COMPARE
304 Stainless Steel 400 Burkle, Inc. 5301-2000 Silo drill sampler, manual operation, SS, 150 cm long, 90 mm screw, 40 mm chamber; 400 mL