Bürkle Dipping Vessels
Ideal for all-layer sampling of liquids
  • Suitable for Explosion Categories IIA, IIB, and IIC
  • Tested for use in Ex protection zone 0
These dipping vessels are ideal for all-layer samples from tanks, tank wagons, water courses, and water purification plants. Easy-flow valves allow liquid to constantly run through the vessel as it is lowered. As soon as the vessel is pulled back up, the valves automatically close.
457,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Burkle 5365-2000 Dipping Vessel, SS, 82 mm; 1000 mL WW-06042-20 Mfr # 5365-2000
COMPARE
304 Stainless Steel
₹457,000.00INR / Each