B&K Precision Isolated Variable AC Power Supply
FEATURES:
  • Power line leakage tester
  • 2 Amp continuous output
  • Displays voltage or current readings
  • Isolated transformer to eliminate shock hazard while servicing ''hot'' chassis equipment
  • Compact, rugged
190,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1655A 150 Volt 3 Amp, Isolated Variable AC Power Supply WW-20017-11 Mfr # 1655A
COMPARE
3 A 150 VAC
₹190,000.00INR / Each