B&K Precision Dual Range DC Power Supplies
Precision power at an economical price
  • Dual range to accommodate applications requiring either a higher voltage or a higher current
  • Low ripple and noise and excellent regulation
  • Constant voltage (CV) and constant current (CC) mode operation with LED indication
  • Two bright 4-digit LED displays for CV and CC modes
  • Automatic recall of last settings on power up
  • RS-232 interface enables remote control via PC
  • Isolated output
  • Overload protection
  • Reverse polarity protection
These power supplies are general purpose dual range DC power sources. Instruments can output higher voltage at a lower current range or more current at a lower voltage range. The dual 4-digit LED displays continuously monitor the output voltage and current. The power supplies can be operated locally from the front panel or remotely through the RS-232 interface. Units provide excellent regulation and low ripple characteristics. The mechanical configuration conserves bench space and allows for easy portability. These power supplies are well-suited for a wide variety of electrical and electronics applications in service shops, engineering labs, production, and laboratories and more.
177,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1737 Dual Range DC Power Supply, 30V/3A, 60V/2A WW-20045-76 Mfr # 1737
COMPARE
0 to 3 A, 0 to 2 A 0 to 30 VDC, 0 to 60 VDC < 1 mVrms
₹177,000.00INR / Each