This product is now discontinued, View Related Products
B&K Precision Digital Function Generators
Produce waveforms at your specified frequencies
  • Adjustable attenuation of 0 to 20 dB
  • Model 26864-52 includes an internal frequency counter up to 30 MHz
These units generate sine, square, triangle, ramp, pulse, TTL and CMOS outputs, and offer variable duty cycle, and variable DC offset. Use coarse and fine tuning controls to accurately display the necessary reading on the five-digit LED display.
Model 26864-50 generates signals between 0.1 Hz to 10 MHz, in eight selectable ranges. This unit provides normal mode, adjustable linear or log sweep, and voltage controlled generator (VCG) functions.
Model 26864-52 generates signals between 0.2 Hz to 20 MHz, in eight selectable ranges. It offers the same features as the 26864-50, plus AM and FM modulation, burst mode and provides a built-in frequency counter up to 30 MHz.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Frequency Range (Hz) Number Of Channels Wave Form
Availability Pricing
B&K Precision 4017A Digital Function Generator, 10 MHz WW-26864-50 Mfr # 4017A
COMPARE
Call For Price
B&K Precision 4040A Digital Function Generator, 20 MHz WW-26864-52 Mfr # 4040A