This product is now discontinued, View Related Products
B&K Precision DC Electronic Load Modules
Choose up to four load modules for your mainframe
  • Modules and mainframes sold separately
The MDL Series is a multi-channel modular programmable electronic load system. Seven different modules of programmable DC loads ranging in power from 200 W to 600 W provide users the flexibility to test a wide range of power sources from multi-output AC/DC power supplies to batteries, fuel cells, and photovoltaic arrays.
The mainframe has four slots that can be configured with any assortment of the modules up to 2400 W (up to 4800 W with mainframe extension). The high-performance electronic load modules of the MDL Series are capable of operating in constant current (CC), constant voltage (CV), constant resistance (CR), constant power (CW), and constant impedance (CZ) mode, which uses DSP technology to simulate non-linear loads and realistic loading behavior.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
B&K Precision MDL200 DC Electronic Load Module, 1-channel, 200 W / 80 V / 40 A WW-20056-21 Mfr # MDL200
COMPARE
DC Electronic Load Module, 1-channel, 200 W / 80 V / 40 A
Call For Price
B&K Precision MDL200 DC Electronic Load Module, 2-channel, 250/50 W / 80 V / 20 A WW-20056-22 Mfr # MDL252
COMPARE
DC Electronic Load Module, 2-channel, 250/50 W / 80 V / 20 A
Call For Price
B&K Precision DC Electronic Load Module, 2-channel, 300 W/300 W / 80 V / 45 A WW-20056-23 Mfr # MDL302
COMPARE
DC Electronic Load Module, 2-channel, 300 W/300 W / 80 V / 45 A
Call For Price
B&K Precision DC Electronic Load, 1-channel, 300 W/500 V/20 A WW-20056-24 Mfr # MDL305
COMPARE
DC Electronic Load, 1-channel, 300 W/500 V/20 A
Call For Price
B&K Precision DC Electronic Load Module, 1-channel, 400 W/80 V/60 A WW-20056-25 Mfr # MDL400
COMPARE
DC Electronic Load Module, 1-channel, 400 W/80 V/60 A
Call For Price
B&K Precision DC Electronic Load Module, 1-channel, 500 W/500 V/30 A WW-20056-26 Mfr # MDL505
COMPARE
DC Electronic Load Module, 1-channel, 500 W/500 V/30 A
Call For Price
B&K Precision DC Electronic Load Module, 1-channel, 600 W/80 V/120 A WW-20056-27 Mfr # MDL600
COMPARE
DC Electronic Load Module, 1-channel, 600 W/80 V/120 A
Call For Price
Showing 1- 7 of 7