B&K Precision 9800 Series Programmable AC Power Sources
Program and perform electronics R&D and precompliance tests from your PC
  • Expansive programmability options include PLD and dimmer simulators, surges, sags, frequency disturbances, and sweeps
  • USB, RS-232, LAN, and GPIB interfaces and LabVIEW™ compatibility support remote control by PC
  • Conveniently trigger programs via front panel, BNC connector on the rear panel, or by PC
  • Measure rms voltage, rms current, peak current, and power during tests
This programmable power source is ideally suited to evaluation of TRIACs and SCRs, transformers, and passive electrical elements. Adjustable phase angle in the dimmer simulation mode is perfect for testing motors and lighting components. The bright vacuum fluorescent display (VFD) shows actual and measured parameters, as well as mode and error indicators.
The front-panel output terminal is a universal socket, which accommodates AC plugs from the US, UK, Canada, China, and Europe, while the rear panel features a terminal block output. Compact housing is 2U rack-mountable.
Safety features include key-lock function and protections against overvoltage, overcurrent, overpower, and overtemperature conditions.
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
B&K Precision 9801 Programmable 300 VA AC Power Source WW-20056-00 Mfr # 9801
COMPARE
Programmable 300 VA AC Power Source
₹710,000.00INR / Each