This product is now discontinued, View Related Products
B&K Precision 9150 Series Single Output Programmable DC Power Supplies
Provides excellent display resolution with fast transient response time (<100 µs)
  • Very low ripple and noise due to linear regulation
  • Programmable via USB and RS-232 using SCPI compatible command set
  • List mode operation for increased throughput
  • Very quiet due to intelligent fan speed control
  • Bench or rack mountable
  • Included software for front panel emulation and simple test sequence generation
  • Digital port with remote inhibit (RI) useful for turning multiple power supplies On/Off simultaneously
  • Over voltage protection
  • Digital Volt Meter and Milliohm Meter
These power supplies are designed for applications in design verification, production, or university labs that require high, clean, and reliable power combined with excellent resolution, accuracy, and fast transient response time.
4 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 9150 5.2V / 60A Programmable DC Power Supply WW-20036-63 Mfr # 9150
COMPARE
0 to 60 A 0 to 5.2 VDC ≤ 4 mVp-p; ≤ 15 mArms
Call For Price
B&K Precision 9151 20V / 27A Programmable DC Power Supply WW-20036-64 Mfr # 9151
COMPARE
0 to 27 A 0 to 20 VDC ≤ 4 mVp-p; ≤ 5 mArms
Call For Price
B&K Precision 9152 30V / 18A Programmable DC Power Supply WW-20036-65 Mfr # 9152
COMPARE
0 to 18 A 0 to 30 VDC ≤ 4 mVp-p; ≤ 5 mArms
Call For Price
B&K Precision 9153 60V / 9A Programmable DC Power Supply WW-20036-66 Mfr # 9153
COMPARE
0 to 9 A 0 to 60 VDC ≤ 5 mVp-p; ≤ 3 mArms