This product is now discontinued, View Related Products
B&K Precision 4075B Series Arbitrary/Function Generators
Versatile high performance with the largest arbitrary memory depth in its class
  • Generate sine waveforms
  • Flash memory can be allocated to one large waveform or up to 49 different waveforms
  • Conveniently add noise to your waveform, directly from the front panel, and precisely adjust the scale of the noise amplitude
  • Fully programmable markers and protected output
  • Includes built-in 10 MHz external reference
  • Integration with WaveXpress™ Waveform Editing software
The 4075B series generators combine the ability to produce nearly any conceivable arbitrary waveform with accuracy and precision, and feature a DDS architecture to provide conventional function generator capabilities. Extensive features such as internal or external AM, FM, and FSK modulation along with versatile sweep capabilities and variable edge pulse generation make these generators suitable for a wide range of applications including electronic design, sensor simulation, and functional test or generation of I/Q modulated signals.
Waveforms can be output in continuous, triggered, gated or burst modes, and units can be remotely controlled using SCPI compliant commands via USB for additional flexibility. Without requiring any other instruments be connected to the computer, the free WaveXpress software allows upload and download of waveforms to and from the AWG, and the testing of actual or created waveforms with the drawing and editing tools.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Frequency Range (Hz) Number Of Channels Wave Form
Availability Pricing
B&K Precision 4075B Single Channel Arbitrary/Function Generator, 30 MHz WW-20033-25 Mfr # 4075B
COMPARE
1 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle
Call For Price
B&K Precision 4078B Dual Channel Arbitrary/Function Generator, 25 MHz WW-20033-26 Mfr # 4078B
COMPARE
2 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle
Call For Price
B&K Precision 4076B Single Channel Arbitrary/Function Generator, 50 MHz WW-20036-60 Mfr # 4076B
COMPARE
1 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle
Call For Price
B&K Precision 4079B Dual Channel Arbitrary/Function Generator, 50 MHz WW-20036-61 Mfr # 4079B
COMPARE
2 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle
Call For Price
B&K Precision 4077B Model , Single Channel Arbitrary/Function Generator, 80 MHz WW-20048-78 Mfr # 4077B
COMPARE
2 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle
Call For Price
B&K Precision 4080B Model , Dual Channel Arbitrary/Function Generator, 80 MHz WW-20048-79 Mfr # 4080B
COMPARE
1 Exponential up and down, gaussian, ramp up/down, noise, sine, sin(x)/x, square, triangle