B&K Precision 1.44 kW Programmable DC Power Supplies
Compact, high density, 1U package provides up to 1.44 kW of power
These power supplies combine fast average command processing times of less than 50 ms, with the flexibility of industry-standard GPIB, LAN, USB & RS485 interfaces making them ideal for ATE applications. Built-in voltage and current meters display settings and output values concurrently, and units provide standard USB & RS485 interfaces (-GL models include GPIB and LAN). Power supplies feature easy-to-configure master/slave mode for series & parallel connection of up to 4 units, OVP, OCP, OPP protection, and the ability to control up to 31 power supplies from one PC via the RS485 interface.
850,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision XLN3640-GL 1.44kW DC Power Supply 36V/40A/USB/GPIB/LAN WW-20035-83 Mfr # XLN3640-GL
COMPARE
0 to 40 A 0 to 36 VDC ≤ 5 mVrms, ≤ 60 mVp-p
₹850,000.00INR / Each