This product is now discontinued, View Related Products

Azlon LDPE Solvent Venting Wash Bottles, 500 mL

Eliminate dripping and sputtering
  • Fill bottles easily with wide-mouth design
  • Easy identification with
  • color-coding
These bottles feature a valve mechanism that automatically vents volatile solvent vapor and eliminates the potentially hazardous problems of solvent drip and sputter. A wide mouth makes bottles easy to fill. Closures are color-coded for easy identification.

Chemical name, formula, hazard information, and instructions are printed on the bottle.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Label Type Material
Availability Pricing
Azlon 506465-0001 LDPE Solvent Venting Wash Bottle, acetone, 500 mL, 5/pk WW-62303-40 Mfr # 506465-0001
COMPARE
500 Acetone Low Density Polyethyene (LDPE)
Call For Price
Azlon 506465-0002 LDPE Solvent Venting Wash Bottle, ethanol, 500 mL, 5/pk WW-62303-50 Mfr # 506465-0002
COMPARE
500 Ethanol Low Density Polyethyene (LDPE)
Call For Price
Azlon 506465-0004 LDPE Solvent Venting Wash Bottle, isopropanol, 500 mL, 5/pk WW-62303-60 Mfr # 506465-0004
COMPARE
500 Isopropanol Low Density Polyethyene (LDPE)
Call For Price
Azlon 506465-0005 LDPE Solvent Venting Wash Bottle, methanol, 500 mL, 5/pk WW-62303-70 Mfr # 506465-0005
COMPARE
500 Methanol Low Density Polyethyene (LDPE)
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat