Ashcroft® Model 1082 and 1084 Pressure Test Gauges
For checking or calibrating other gauges or in applications where high accuracy is required
  • MicroSpan™ adjustment for ease in span calibration
Gauges are factory calibrated to ensure accuracy. To determine the right gauge for your application, choose the Bourdon tube and process connection material according to the conditions and medium to which the gauge will be exposed. Then choose your desired range and accuracy. For added safety, the maximum continuous pressure applied to the gauge should not exceed 75% of the maximum pressure of the gauge. All gauges have a 1/4'' NPT(M) process connection. The 3'' gauges include a soft carrying case; order hard carrying case 68030-90 for 4.5'' and 6'' dial gauges.
39,100.00 - ₹77,700.00INR / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 30 to 0" Hg WW-68030-01 Mfr # 45-1082-AS-02L-30IHGV
COMPARE
4.5\" Test Pressure Gauge 30-0\"Hg, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 15 psi WW-68030-03 Mfr # 45-1082-AS-02L-15#
COMPARE
4.5\" Test Pressure Gauge 0-15 psi, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 30 psi WW-68030-04 Mfr # 45-1082-AS-02L-30#
COMPARE
4.5\" Test Pressure Gauge 0-30 psi, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 60 psi WW-68030-05 Mfr # 45-1082-AS-02L-60#
COMPARE
4.5\" Test Pressure Gauge 0-60 psi, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 100 psi WW-68030-06 Mfr # 45-1082-AS-02L-100#
COMPARE
4.5\" Test Pressure Gauge 0-100 psi, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 200 psi WW-68030-07 Mfr # 45-1082-AS-02L-200#
COMPARE
4.5\" Test Pressure Gauge 0-200 psi, 1/4\" NPT Lower
₹50,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Monel 400 Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 15 psi WW-68031-03 Mfr # 45-1082-PS-02L-15#
COMPARE
Test Gauge 0.25% Accuracy, Monel Bourdon Tube, 4 1/2\" Dialm 0 to 15 psi
₹77,700.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Monel 400 Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 30 psi WW-68031-04 Mfr # 45-1082-PS-02L-30#
COMPARE
4.5\" Monel Test Pressure Gauge 0-30 psi, 1/4\" NPT Lower
₹77,700.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Monel 400 Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 60 psi WW-68031-05 Mfr # 45-1082-PS-02L-60#
COMPARE
Test Gauge 0.25% Accuracy, Monel Bourdon Tube, 4 1/2\" Dial, 0 to 60 psi
₹77,700.00INR / Each
Ashcroft Test Gauge, Model 1082, 4.5" Dial, Monel 400 Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 1000 psi WW-68031-09 Mfr # 45-1082-PS-02L-1000#
COMPARE
4.5\" Monel Test Pressure Gauge 0-1000 psi, 1/4\" NPT Lower
₹77,700.00INR / Each
Ashcroft Test Gauge, Model 1082, 6" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 30 to 0" Hg WW-68032-01 Mfr # 60-1082-AS-02L-30IHGV
COMPARE
Test Gauge 0.25% Accuracy, Bronze Bourdon Tube, 6\" Dial, 30 to 0\" Hg
₹60,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 6" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 30" Hg to 30 psi WW-68032-02 Mfr # 60-1082-AS-02L-30IHGV/30#
COMPARE
6\" Test Pressure Gauge 30\"Hg-30psi, 1/4\" NPT Lower
₹60,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 6" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 30 psi WW-68032-04 Mfr # 60-1082-AS-02L-30#
COMPARE
Test Gauge 0.25% Accuracy, Bronze Bourdon Tube, 6\" Dial, 0 to 30 psi
₹60,400.00INR / Each
Ashcroft Test Gauge, Model 1082, 6" Dial, Bronze/Brass Bourdon Tube, 1/4" NPT(M) Lower Mount, 0 to 200 psi WW-68032-07 Mfr # 60-1082-AS-02L-200#
COMPARE
Test Gauge 0.25% Accuracy, Bronze Bourdon Tube, 6\" Dial, 0 to 200 psi
₹60,400.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 30 to 0" Hg WW-68034-01 Mfr # 30-1084-S-02L-30IHGV
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 30 to 0\" Hg
₹39,100.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 30" Hg to 30 psi WW-68034-02 Mfr # 30-1084-S-02L-30IHGV/30#
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 30\" Hg to 30 psi
₹39,100.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 0 to 15 psi WW-68034-03 Mfr # 30-1084-S-02L-15#
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 0 to 15 psi
₹39,100.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 0 to 30 psi WW-68034-04 Mfr # 30-1084-S-02L-30#
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 0 to 30 psi
₹39,100.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 0 to 60 psi WW-68034-05 Mfr # 30-1084-S-02L-60#
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 0 to 60 psi
₹39,100.00INR / Each
Ashcroft Test Gauge, Model 1084, 3" Dial, 316L Stainless Steel, 1/4" NPT(M) Lower Mount, 0 to 100 psi WW-68034-06 Mfr # 30-1084-S-02L-100#
COMPARE
Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3\" Dial, 0 to 100 psi
₹39,100.00INR / Each
Showing 1 - 20 of 21