Ashcroft® Model 1188 Low-Pressure/Vacuum Bellows Gauges
The Ashcroft® 1188 bellows gauge is used when an ASME style low pressure gauge is required
  • A variety of wetted parts compatable with many process medias
  • Ideal product solution for many installations
  • Overpressure capability up to 5x full scale range standard
Its highly sensitive system and standard product features are available to meet many process applications. Available with diaphragm seals, this is an ideal product instrument solution for many installation points. With ranges as low as 10 inches of water vacuum, a variety of wetted parts and case materials and solid front case designs, these low pressure gauges are generally used in applications such as oil & gas, chemical/petrochemical plants, water/wastewater pressure control, and equipment skids.
104,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (in WC) Min Pressure (in WC)
Availability Pricing
Ashcroft 4.5" Low-Pressure Brass Bellows Gauge, 0 to 30" WC WW-68427-50 Mfr # 45-1188-AS-04L-30IW/ZSI
COMPARE
30 0
₹104,000.00INR / Each