Ashcroft LTD Series Temperature Switches
Dual setpoints for added process control
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—meet NEMA 4, 4X, and IP66 requirements
Dual set point, fixed deadband, SPDT switches feature epoxy-coated aluminum systems for reliable performance. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration, velocity, or pressure. Remote-mount units have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft LTD Temperature Switch Remote Mount, 15' Capillary, 75/205°F, 2 Ind. Setpoints w/ Adapter WW-39012-04 Mfr # LTD N4 GG 15-A7-030 XBX 75/205F
COMPARE
Temperature Switch Remote Mount, 15' Capillary, 75/205°F, 2 Ind. Setpoints w/ Adapter
₹161,000.00INR / Each