Ashcroft® Stainless Steel Industrial Process Gauge
Gauge features solid front safety case for harsh conditions
  • 0.5% accuracy complies with ASME B40.1 Grade 2A
  • Micrometer adjustable pointer
  • 316L stainless steel Bourdon tube and socket
Industries for these rugged process gauges include refineries, petrochemical, chemical, wastewater, and pulp and paper. Bourdon tube is all welded for safety and longer life. Range is vacuum to 20,000 psi.
Heavy- duty gauges are available filled and unfilled; liquid fill is glycerin. Gauge ring features cam lock and fiberglass reinforced thermoplastic; movement is 300 series stainless steel. Gauge’s window is glass. Proof pressure of these industrial gauges is 130% of full scale.
13,400.00 - ₹16,500.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Accuracy Description
Availability Pricing
Ashcroft 4.5" Industrial Process Gauge 30 to 0" Hg WW-68337-00 Mfr # 45-1259-SD-04L-30IHGV
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 30 to 0\" Hg
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, 30" Hg to 30 psi WW-68337-02 Mfr # 45-1259-SD-04L-30IHGV/30#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, 30\" Hg to 30 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 15 psi WW-68337-04 Mfr # 45-1259-SD-04L-15#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 15 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 30 psi WW-68337-06 Mfr # 45-1259-SD-04L-30#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 30 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 60 psi WW-68337-08 Mfr # 45-1259-SD-04L-60#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 60 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 100 psi WW-68337-10 Mfr # 45-1259-SD-04L-100#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 100 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 200 psi WW-68337-12 Mfr # 45-1259-SD-04L-200#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 200 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 400 psi WW-68337-14 Mfr # 45-1259-SD-04L-400#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 400 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 600 psi WW-68337-16 Mfr # 45-1259-SD-04L-600#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 600 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge 0 to 1000 psi WW-68337-18 Mfr # 45-1259-SD-04L-1000#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge 0 to 1000 psi
₹13,400.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 30 to 0" Hg WW-68337-28 Mfr # 45-1259-SL-04L-30IHGV
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 30 to 0\" Hg
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 15 psi WW-68337-32 Mfr # 45-1259-SL-04L-15#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 15 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 30 psi WW-68337-34 Mfr # 45-1259-SL-04L-30#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 30 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 60 psi WW-68337-36 Mfr # 45-1259-SL-04L-60#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 60 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 100 psi WW-68337-38 Mfr # 45-1259-SL-04L-100#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 100 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 200 psi WW-68337-40 Mfr # 45-1259-SL-04L-200#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 200 psi
₹16,500.00INR / Each
Ashcroft 4.5" Industrial Process Gauge, Glycerin Filled, 0 to 400 psi WW-68337-42 Mfr # 45-1259-SL-04L-400#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 4.5\" Industrial Process Gauge, Glycerin Filled, 0 to 400 psi
₹16,500.00INR / Each
Showing 1- 17 of 17