This product is now discontinued, View Related Products
Ashcroft D-Series Differential Pressure Switches
Differential pressure switch designed for your hazardous applications
Watertight differential pressure switches are ideal for industrial and OEM applications and have fixed set point and deadband. Pressure switche enclosures meet NEMA 3, 4, 4X, 13, and IP66 requirements.
Explosion proof differential pressure switch are designed for hazardous process and industrial applications. Switches have fixed setpoint and deadband. Explosion-proof enclosure meets Div. 1 and 2, MENA 7, 9, and IP66 requirements. These switches meet UL, CSA, ATEX and IECEx approval for hazardous locations.
Call For Price
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft D Series Explosion Proof Pressure Switch, 30" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-00 Mfr # D724B30IWD
COMPARE
Explosion Proof Pressure Switch, 30\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 60" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-01 Mfr # D724B60IWD
COMPARE
Explosion Proof Pressure Switch, 60\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 100" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-02 Mfr # D724B100IWD
COMPARE
Explosion Proof Pressure Switch, 100\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 150" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-03 Mfr # D724B150IWD
COMPARE
Explosion Proof Pressure Switch, 150\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 15 PSID, 1/4" NPT(F); Single Set Point WW-68361-04 Mfr # D724B15#D
COMPARE
Explosion Proof Pressure Switch, 15 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 30 PSID, 1/4" NPT(F); Single Set Point WW-68361-05 Mfr # D724B30#D
COMPARE
Explosion Proof Pressure Switch, 30 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof ressure Switch, 60 PSID, 1/4" NPT(F); Single Set Point WW-68361-06 Mfr # D724B60#D
COMPARE
Explosion Proof ressure Switch, 60 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 100 PSID, 1/4" NPT(F); Single Set Point WW-68361-07 Mfr # D724B100#D
COMPARE
Explosion Proof Pressure Switch, 100 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof ressure Switch, 200 PSID, 1/4" NPT(F); Single Set Point WW-68361-08 Mfr # D724B200#D
COMPARE
Explosion Proof ressure Switch, 200 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 400 PSID, 1/4" NPT(F); Single Set Point WW-68361-09 Mfr # D724B400#D
COMPARE
Explosion Proof Pressure Switch, 400 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Explosion Proof Pressure Switch, 600 PSID, 1/4" NPT(F); Single Set Point WW-68361-10 Mfr # D724B600#D
COMPARE
Explosion Proof Pressure Switch, 600 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 30" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-11 Mfr # D424B30IWD
COMPARE
Pressure Switch, 30\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 60" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-12 Mfr # D424B60IWD
COMPARE
Pressure Switch, 60\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 100" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-13 Mfr # D424B100IWD
COMPARE
Pressure Switch, 100\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 150" WC Diff, 1/4" NPT(F); Single Set Point WW-68361-14 Mfr # D424B150IWD
COMPARE
Pressure Switch, 150\" WC Diff, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 15 PSID, 1/4" NPT(F); Single Set Point WW-68361-15 Mfr # D424B15#D
COMPARE
Pressure Switch, 15 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 30 PSID, 1/4" NPT(F); Single Set Point WW-68361-16 Mfr # D424B30#D
COMPARE
Pressure Switch, 30 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 60 PSID, 1/4" NPT(F); Single Set Point WW-68361-17 Mfr # D424B60#D
COMPARE
Pressure Switch, 60 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 100 PSID, 1/4" NPT(F); Single Set Point WW-68361-18 Mfr # D424B100#D
COMPARE
Pressure Switch, 100 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Ashcroft D Series Pressure Switch, 200 PSID, 1/4" NPT(F); Single Set Point WW-68361-19 Mfr # D424B200#D
COMPARE
Pressure Switch, 200 PSID, 1/4\" NPT(F); Single Set Point
Call For Price
Showing 1 - 20 of 22