Ashcroft® CXLdp Low Pressure Transducers
Provides high-accuracy in energy management and comfort control
  • Very small footprint
  • ±0.8 and ±0.4% accuracy versions available
  • TruAccuracy™- Terminal Point Accuracy method includes non-linearity, hysteresis, non-repeatability, zero offset and span setting errors
  • Rugged ABS package capable of DIN rail or panel mounting
  • LED power status indicator
  • Detachable Euro style terminal block
  • Unidirectional and Bidirectional ranges
These differential pressure transducers feature a micro-machined silicon capacitive sensor with extraordinary sensitivity, repeatability and stability of output. The small physical size requires less space in a panel, and can be DIN rail mounted for faster installation and neater appearance. Detachable terminal block reduces wiring errors and field wiring time. A green LED indicates the transducer is powered and functioning and also helps locate the transducer in dark ducts, unlit ceilings, and electrical enclosures. Also, brightness varies with pressure. The conduit/plenum mounting kit comes standard.
38,300.00 - ₹49,700.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (in WC)
Availability Pricing
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.40% of Span, 1/4" M Barb Connection, 4-20 mA, 1.00" Water Column WW-68333-24 Mfr # CX-4-MB2-42-1IW-XAH
COMPARE
±0.4% full-scale 1
₹49,700.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.40% of Span, 1/4" M Barb Connection, 4-20 mA, 2.00" Water Column WW-68333-26 Mfr # CX-4-MB2-42-2IW-XAH
COMPARE
±0.4% full-scale 2
₹49,700.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.40% of Span, 1/4" M Barb Connection, 4-20 mA, 2.50" Water Column WW-68333-28 Mfr # CX-4-MB2-42-2P5IW-XAH
COMPARE
±0.4% full-scale 2.5
₹49,700.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.40% of Span, 1/4" M Barb Connection, 4-20 mA, ±0.10" Water Column WW-68333-38 Mfr # CX-4-MB2-42-P1IWL-XAH
COMPARE
±0.4% full-scale 0.1
₹49,700.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.40% of Span, 1/4" M Barb Connection, 4-20 mA, ±1.00" Water Column WW-68333-44 Mfr # CX-4-MB2-42-1IWL-XAH
COMPARE
±0.4% full-scale 1
₹49,700.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.80% of Span, 1/4" M Barb Connection, 4-20 mA, 0.25" Water Column WW-68333-52 Mfr # CX-8-MB2-42-P25IW-XAH
COMPARE
±0.8% full-scale 0.25
₹38,300.00INR / Each
Ashcroft CXLdp Differential Pressure Transducer with Plenum/Conduit Kit, ±0.80% of Span, 1/4" M Barb Connection, 4-20 mA, 25.00" Water Column WW-68333-68 Mfr # CX-8-MB2-42-25IW-XAH
COMPARE
±0.8% full-scale 25
₹38,300.00INR / Each
Showing 1- 7 of 7