This product is now discontinued, View Related Products
Arioso Power Scholar Water Purification Systems
Single unit delivers pure or ultrapure water with one touch
  • Touch screen and plain language menus make setup easy
  • Dedicated pure and ultrapure water dispense icons on main screen simplify operation
  • Open front door to quickly change treatment cartridges
This combination reverse osmosis (RO) and ultrapure (UP) treatment water purification system provides a complete solution to your laboratory-grade water requirements. Pure water, produced by the RO component, is suitable for general laboratory applications, such as reagent and media preparation, glassware rinsing, and autoclaving. Ultrapure water, produced when both RO and UP components are used in conjunction, is necessary for sensitive analytical applications, such as high-performance liquid chromatography (HPLC), gas chromatography (GC), mass spectroscopy (MS) and other critical uses.
Compact unit fits on the bench top near your workspace for access at point of use. Convenient tap water supply feed works in any lab.
Scholar Models feature either a 0.2 µm or a pyrogen-free final filter, and are available with three different levels of production (15, 25, or 35 L/hr).
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description Max Production (L/hour) Power (VAC)
Availability Pricing
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 15 L/hr, 220 VAC WW-67000-00 Mfr # AP1-S-220
COMPARE
Water Purification System with 0.2 µm Filter, 15 L/hr, 220 VAC 15 220
Call For Price
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 15 L/hr, 120 VAC WW-67000-01 Mfr # AP1-S-120
COMPARE
Water Purification System with 0.2 µm Filter, 15 L/hr, 120 VAC 15 120
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 15 L/hr, 220 VAC WW-67000-02 Mfr # AP1-SF-220
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 15 L/hr, 220 VAC 15 220
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 15 L/hr, 120 VAC WW-67000-03 Mfr # AP1-SF-120
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 15 L/hr, 120 VAC 15 120
Call For Price
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 25 L/hr, 220 VAC WW-67000-13 Mfr # AP2-S-220
COMPARE
Water Purification System with 0.2 µm Filter, 25 L/hr, 220 VAC 25 220
Call For Price
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 25 L/hr, 120 VAC WW-67000-14 Mfr # AP2-S-120
COMPARE
Water Purification System with 0.2 µm Filter, 25 L/hr, 120 VAC 25 120
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 25 L/hr, 220 VAC WW-67000-15 Mfr # AP2-SF-220
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 25 L/hr, 220 VAC 25 220
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 25 L/hr, 120 VAC WW-67000-16 Mfr # AP2-SF-120
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 25 L/hr, 120 VAC 25 120
Call For Price
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 35 L/hr, 220 VAC WW-67000-32 Mfr # AP3-S-220
COMPARE
Water Purification System with 0.2 µm Filter, 35 L/hr, 220 VAC 35 220
Call For Price
Arioso Power Scholar Water Purification System with 0.2 µm Filter, 35 L/hr, 120 VAC WW-67000-33 Mfr # AP3-S-120
COMPARE
Water Purification System with 0.2 µm Filter, 35 L/hr, 120 VAC 35 120
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 35 L/hr, 220 VAC WW-67000-34 Mfr # AP3-SF-220
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 35 L/hr, 220 VAC 35 220
Call For Price
Arioso Power Scholar Water Purification System w/ Pyrogen-Free Filter, 35 L/hr, 120 VAC WW-67000-35 Mfr # AP3-SF-120
COMPARE
Water Purification System w/ Pyrogen-Free Filter, 35 L/hr, 120 VAC 35 120
Call For Price
Showing 1- 12 of 12