This product is now discontinued, View Related Products
Argos Technologies® ThinSpin™ Low-Profile Stirrer
Use and store in tight spaces
  • Sleek, low-profile design is ideal for use in fume hoods and cabinets
  • Change direction from clockwise to counterclockwise with the push of a button
  • Program and stir from 15 to 1500 rpm in 15-rpm increments
This ultrathin, lightweight magnetic stirrer can be programmed to mix up to 500 mL at speeds from 15 to 1500 rpm, in increments of 15 rpm. Alternate spin direction from clockwise or counterclockwise by pressing the reverse key.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
100 to 240 Thinspin Low Profile Magnetic Stirrer, 100-240 VAC; US Adaptor
Call For Price
COMPARE
100 to 240 Thinspin Low Profile Magnetic Stirrer, 100-240 VAC; International Adaptor