Cole-Parmer® Essentials Reusable Inoculating Needles
Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
1,600.00 - ₹8,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Teasing Needle, 6\", Straight, 19 Guage, White; Each
₹3,200.00INR / Each
COMPARE
Teasing Needle, 6\", Angled, 19 Guage, Blue; Each
₹3,200.00INR / Each
COMPARE
Inoculating Needles, 3\", 25 Gauge; 12/Pk
₹1,600.00INR / Pkg of 12
COMPARE
Inoculating Needle, Micro, Clear; 5/Pk
₹8,700.00INR / Pkg of 5