This product is now discontinued, View Related Products
Argos Technologies® PolarSafe® Vertical/Chest Freezer Racks for 50-Cell 1.5 mL Microtube Storage Boxes
Easily place and retrieve racks using sturdy dual handles
  • Each compartment holds two 50-cell 1.5 mL microtube storage boxes
  • Included locking rods securely holds boxes in place
  • Strudy corrosion-resistant stainless steel construction for long-term use
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
26 29 15/16
Call For Price
COMPARE
24 27 11/16
Call For Price
COMPARE
22 25 3/8
Call For Price
COMPARE
20 23 1/16
Call For Price
COMPARE
18 20 3/4
Call For Price
COMPARE
16 18 7/16
Call For Price
COMPARE
14 16 1/8
Call For Price
COMPARE
12 13 7/8
Call For Price
COMPARE
10 11 9/16
Call For Price
COMPARE
8 9 1/4
Call For Price
COMPARE
6 6 15/16
Call For Price
Showing 1- 11 of 11