This product is now discontinued, View Related Products
Argos Technologies® PolarSafe® Vertical/Chest Aluminum Racks for Mini 2" Boxes
Aluminum racks offer better temperature uniformity in vapor phase in liquid nitrogen tanks
  • Lightweight aluminum construction for easier handling
  • Use with a wide variety of chest freezers and liquid nitrogen tanks
  • Accommodates mini 2" cardboard or plastic boxes
  • Locking rod keeps sample storage boxes securely in the rack
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
9 boxes 19 13/16
Call For Price
COMPARE
10 boxes 22
Call For Price
COMPARE
11 boxes 24 3/16
Call For Price
COMPARE
12 boxes 26 3/8
Call For Price
COMPARE
13 boxes 28 1/4
Call For Price
COMPARE
14 boxes 30 13/16
Call For Price
COMPARE
15 boxes 33
Call For Price
Showing 1- 7 of 7