This product is now discontinued, View Related Products
Argos Technologies® PolarSafe® Ice Buckets and Pans
Perfectly suited for use with ice, dry ice, and slurry ice
  • Lightweight and more economical than solid plastic models
  • Provide excellent insulating properties to help keep temperature sensitive samples safely chilled
  • Designed for steady and sturdy benchtop use and will not leave condensation
Buckets and pans are cool containers made of expanded polystyrene that are an economical alternative to polyurethane or other solid-plastic versions. Built in handles and lips make them easy to carry.
3,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Color Description
Availability Pricing
COMPARE
2.5 Black Ice Bucket with Lid, 2.5 L, Polystyrene, Black; Each
Call For Price
COMPARE
5 Blue Ice Bucket with Lid, 5 L, Polystyrene, Blue; Each
Call For Price
COMPARE
1 Black Ice Pan, 1 L, Polystyrene, Black; Each
Call For Price
COMPARE
4 Blue Ice Pan, 4 L, Polystyrene, Blue; Each
₹3,400.00INR / Each
COMPARE
7.5 Green Ice Pan, 7.5 L, Polystyrene, Green; Each
Call For Price
COMPARE
5 Black Ice Bucket with Lid, 5 L, Polystyrene, Black; Each