This product is now discontinued, View Related Products
Argos Technologies® Nova™ Microcinerator Sterilizer Accessories
Enhance the versitility of your Nova® Microcinerator Loop and Needle Sterilizer
The slide dryer is a safe and easy way to use heat already generated by the incinerator to dry slides. This is ideal for dissolving epithelium using KOH mycology technique.
The loop rest protects loop handles from overheating or melting. This accessories fits any loop and attaches easily to the incinerators.
The heating element of your Nova® Microcinerator Loop and Needle Sterilizer will last approximatley 6 months depending on usage. Easily and quikly replace these heating elements to keep your incinerator up and running. Model 04397-12 is for the 120V units and 04397-16 is for the 220V powered incinerators.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Microcinerator Replacement Heating Element; 120 VAC
Call For Price
COMPARE
Microcinerator Slide Dryer, 3-Place
Call For Price
COMPARE
Microcinerator Loop Rest, Aluminum
Call For Price
COMPARE
Microcinerator Replacement Heating Element; 220 VAC
Call For Price
COMPARE
Microcinerator Replacement Fuse, 2.5 Amp; 120 VAC
Call For Price
COMPARE
Microcinerator Replacement Fuse, 1.25 Amp; 220 VAC
Call For Price
COMPARE
Microcinerator Replacement Heating Element; 240 VAC
Call For Price
Showing 1- 7 of 7