Argos Technologies® Motorized Pestle Mixer
Quickly homogenize cells, plant tissue, and DNA pellets within microtube vials
  • Free of DNAse, RNAse, and pyrogens to eliminate sample contamination
Use the battery-operated motor mixer with pestles and microtubes to drastically reduce hand fatigue when mixing multiple samples. Cell tissue and pellets are completely homogenized within seconds by means of high-speed vortexing action. Order pestles and tubes separately. The replacement rubber adapter, model 44468-26, is the adapter only, and is only compatible with the 44468-25 pestle mixer. This adapter atteches the motorized mixer to the pestle connector.
1,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Battery-Operated Pestle Motor Mixer; 1 each
Call For Price
COMPARE
Replacement Pestle/Mixer Adapter for Motor; 1 each
₹1,000.00INR / Each