This product is now discontinued, View Related Products
Argos Technologies® Interchangeable Inserts for Modular Pipetting Workstation
  • Interchangeable, inserts can also be used as stand-alone racks
Color-coded polypropylene inserts accept tubes with sizes ranging from 0.5 to 50 mL. Easy-release tabs allow you to quickly change out the inserts to customize the rack to your applications. Each tube cavity is numbered for quick identification. Use the pipetting basin insert to hold up to 25 mL of reagent or solution.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Color Tubes Held Description
Availability Pricing
COMPARE
Red 12 Pipetting Work Station Inserts, 12x0.5mL, Red;5/Pk
Call For Price
COMPARE
Purple 12 Pipetting Work Station Inserts, 12x1.5mL, Purple; 5/Pk
Call For Price
COMPARE
Green 7 Pipetting Work Station Inserts, 7x5mL, Green; 5/Pk
Call For Price
COMPARE
Blue 4 Pipetting Work Station Inserts, 4x15mL, Blue; 5/Pk
Call For Price
COMPARE
Orange 3 Pipetting Work Station Inserts, 3x50mL, Orange; 5/Pk
Call For Price
COMPARE
Yellow 36 Pipetting Work Station Inserts, 36xPCR, Yellow; 5/Pk
Call For Price
COMPARE
Red Pipetting Work Station Inserts ,V-Shaped Basin; 50/pk
Call For Price
Showing 1- 7 of 7