Cole-Parmer® Essentials L-Shaped Cell Spreaders, Sterile
Spread bacterial cultures evenly over agar in petri dishes
  • Sterile to prevent contamination risk
  • L-shape allows application over the entire dish or plate surface
  • Curved end protects agar
Apply cultures over an entire petri dish or plate with these sterile polypropylene cell spreaders. This L-shaped tool has a smooth spreading surface with a slight upward return to reduce the risk of damaging the agar.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these cell spreaders are radiation sterilized (SAL 10-6), DNase/RNase free, nonpyrogenic, and noncytotoxic. Available individually wrapped or bulk packed.
3,800.00 - ₹8,000.00INR / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (mm) Sterile Description
Availability Pricing
COMPARE
145 Yes Cell Spreaders, Sterile, Polypropylene, 50 bags of 10; 500/Cs
₹8,000.00INR / Case of 500
COMPARE
145 Yes L-Shaped Cell Spreaders, Sterile, Polypropylene, Individually Wrapped; 100/Cs
₹3,800.00INR / Case of 100