This product is now discontinued, View Related Products
Ansell Chemical-Resistant PVA Gloves
Extreme resistance to strong solvents, including aromatics, aliphatics, and chlorinated solvents
Poly vinyl alcohol (PVA) gloves are nearly inert to strong solvents, including aromatics, aliphatics, and chlorinated solvents—chemicals which quickly deteriorate natural rubber, neoprene, and PVC gloves. With PVA gloves, there is little absorption to cause swelling and little extraction to cause shrinking. PVA gloves weigh about half as much as neoprene-coated gauntlets, so they provide a high level of comfort. PVA also offers good resistance to snags, punctures, abrasion, and cuts.
Note: PVA coating is water-soluble. Do not use in water or water-based solutions.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Length (in) Size
Availability Pricing
Ansell 214366 Chemical-Resistant PVA Gloves, 12"L, Size 8, 12 pairs/pk WW-09715-01 Mfr # 214366
COMPARE
12 Medium (8-9)
Call For Price
Ansell 214246 Chemical-Resistant PVA Gloves, 12"L, Size 9, 12 pair/pk WW-09715-02 Mfr # 214246
COMPARE
12 Medium (8-9)
Call For Price
Ansell 214244 Chemical-Resistant PVA Gloves, 12"L, Size 10, 12 pair/pk WW-09715-03 Mfr # 214244
COMPARE
12 Large (9-10)
Call For Price
Ansell 214306 Chemical-Resistant PVA Gloves, 14"L, Size 9, 12 pair/pk WW-09715-04 Mfr # 214306
COMPARE
14 9
Call For Price
Ansell 214308 Chemical-Resistant PVA Gloves, 14"L, Size 10, 12 pair/pk WW-09715-05 Mfr # 214308
COMPARE
14 10