This product is now discontinued, View Related Products
Animal-Derived Ingredient Free Sanitary Adapters
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow, no particulate build-up and easy cleaning. Made from animal-derived ingredient-free polypropylene and PVDF, these adapters may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Value Plastics Mini sanitary end cap, 3/4", animal-free polypropylene, pack of 10 WW-31513-90 Mfr # SFMNC-6005
COMPARE
Polypropylene
Call For Price
Value Plastics Sanitary end cap, 1-1/2", animal-free polypropylene, pack of 10 WW-31513-91 Mfr # SFMXC-6005
COMPARE
Polypropylene