Cole-Parmer­­® Essentials Aluminum Weighing Dishes - Smooth Wall
Facilitates easy stacking and dispensing
  • Disposable
  • Can be used in a wide variety of applications
These smooth-sided, flat bottom, multipurpose "utility" dishes are ideal for storage, dispensing, general purpose sampling, and testing. Dish contour facilitates stacking and dispensing. May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical. The dishes are oil-free and non-coated.
3,500.00 - ₹75,100.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Diameter (in) Height (in)
Availability Pricing
COMPARE
80 2 7/8 11/16
₹3,500.00INR / Pkg of 100
COMPARE
80 2 7/8 11/16
₹22,900.00INR / Case of 1000
COMPARE
150 4 3/4
₹75,100.00INR / Case of 1000
COMPARE
150 4 3/4
₹8,800.00INR / Pkg of 100