Alcohol Burner With Ground Glass Cap
Ideal for drying slides
  • Can use with isopropyl or denatured ethyl alcohol
  • Low-heat flame
Manufactured from soda-lime glass, this burner is designed for use with isopropyl or denatured ethyl alcohol. The low-heat flame is well suited for microscopy purposes.
440.00 - ₹8,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gas Type Height (in) Description
Availability Pricing
DWK Life Sciences (Wheaton) 237070 Alcohol Burner, 100 ml Capacity, 4" H x 3" Dia WW-48585-84 Mfr # 237070
COMPARE
Alcohol 4 Alcohol Burner, 100 ml Capacity, 4\" H x 3\" Dia
₹8,800.00INR / Each
DWK Life Sciences (Wheaton) 237071 Replacement Wick for Alcohol Burner, 6" L x 1/4" Dia WW-48585-86 Mfr # 237071
COMPARE
Replacement Wick for Alcohol Burner, 6\" L x 1/4\" Dia
₹440.00INR / Each