Cole-Parmer® Air Cadet® PRO Diaphragm Pumps
Exceptional performance for the price!
  • General purpose pumps are great for filtration or vacuum drying
  • At 45 dB(A) it's quieter than a conversation
  • Long service intervals reduce maintenance costs
Designed for general laboratory applications, these pumps will give you thousands of hours of worry-free operation. Max vacuum of 29.8 in Hg. Pumps with gas ballast will allow purging of any contaminants that may condense in the pump head.
A standard Air Cadet PRO will work for all your basic vacuum needs including filtration and degassing. For demanding applications requiring solvents or corrosive chemicals use the chemical duty Air Cadet PRO
438,000.00 - ₹634,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Power (VAC) Wetted Parts
Availability Pricing
COMPARE
0.6 115 Aluminum, Viton®, Polyamide, PVC
₹438,000.00INR / Each
COMPARE
0.6 115 PTFE, Kalrez®, ETFE
₹634,000.00INR / Each
COMPARE
0.6 230 Aluminum, Viton®, Polyamide, PVC
₹438,000.00INR / Each
COMPARE
0.6 230 PTFE, Kalrez®, ETFE
₹634,000.00INR / Each