This product is now discontinued, View Related Products
Air Admiral Aspiration System
Compact, lightweight portable aspiration system
  • Dial vacuum gauge and regulator to monitor and adjust vacuum level
  • Liquid trap to avoid ingestion of fluids into the pump
  • Automatic flow shut-off when full
Pump features an autoclavable collection receiver made of polypropylene, a hydrophobic inline filter to prevent introduction of the aspirate into the pump and automatic shutoff prevent overflow of the aspirate to protect you and the pump from exposure to the media. The pump also features a vacuum regulator to control the flow of liquid. Recommended for aspirating aqueous solutions including buffers, but not for acidic, basic, or organic vapors and gases. 70202-30 and 79202-35 suitable for bleach solutions.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
Call For Price
COMPARE
230