Advantec Three-Piece 37 mm Monitors for Microbial Analysis
Ideal for bacterial, yeast, mold and other contamination analysis Use 3-piece monitor for aerosol analysis
Pre-assembled 3-piece units contain a built-in membrane filter and a support pad. Monitors are suitable for field collection and transportation of samples as well as analysis in the laboratory. Samples collected on the press-fit membrane can then be cultured in place or removed for further analysis. Individually packaged sets are pre-sterilized.
21,500.00INR / Pkg of 50
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec Three-piece monitor, plain surface, 0.80 um, 50/pack WW-02935-06 Mfr # 37AS380AN
COMPARE
Three-piece monitor, plain surface, 0.80 um, 50/pack
₹21,500.00INR / Pkg of 50