ADAM® LBX Latitude Compact Industrial Bench Scales
Quick and accurate weighing with a portable, compact design
  • Large backlit LCD for quick reading of results in any lighting conditions
  • Never lose power again—rechargeable battery lasts up to 240 hours on a single charge
  • Designed with large, removable stainless steel pan for easy cleaning
  • Constructed with industrial strength housing and sealed keypad for protection in robust environments
These industrial bench scales streamline basic weighing tasks with simple key operation for quick set-up and operation via the easy-to-navigate menu. Scales offer five weighing units and perform weighing, percentage weighing, parts counting, and checkweighing operations. The LBX series scales feature three colored LEDs to indicate under, over, and within limit samples. Advanced features also include hold function to freeze the display without losing results, programmable auto power-off to save battery life, and battery level indicator to clearly indicate when recharging is required.
55,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Readability (g) Description
Availability Pricing
Adam Equipment LBX 12 Latitude Compact Industrial Bench Scale; 12 kg x 2 g WW-11801-81 Mfr # LBX 12
COMPARE
12 2 LBX 12 Latitude Compact Industrial Bench Scale; 12 kg x 2 g
₹55,800.00INR / Each
Adam Equipment LBX 3 Latitude Compact Industrial Bench Scale; 3 kg x 0.5 g WW-11801-82 Mfr # LBX 3
COMPARE
3 0.5 LBX 3 Latitude Compact Industrial Bench Scale; 3 kg x 0.5 g
₹55,800.00INR / Each
Adam Equipment LBX 30 Latitude Compact Industrial Bench Scale; 30 kg x 5 g WW-11801-83 Mfr # LBX 30
COMPARE
30 5 LBX 30 Latitude Compact Industrial Bench Scale; 30 kg x 5 g
₹55,800.00INR / Each
Adam Equipment LBX 6 Latitude Compact Industrial Bench Scale; 6 kg x 1 g WW-11801-84 Mfr # LBX 6
COMPARE
6 1 LBX 6 Latitude Compact Industrial Bench Scale; 6 kg x 1 g
₹55,800.00INR / Each