This product is now discontinued, View Related Products
Adam Equipment Nimbus Analytical Balances
Reliable performance in a compact, rugged design
  • All-metal housing ensures rugged, durable protection and stability
  • Glass doors disassemble for thorough cleaning
  • GLP-compliant printouts via USB or RS-232 connection
  • Below-balance weighing hook
  • Date and time stored in memory
  • Large, backlit display and intuitive keypad
Optimized connections boost data collection capabilities and lead to exceptional information transmission. USB and RS-232 interfaces are standard, while an additional interface allows use of an optional remote display. Whether performing basic tasks such as printing data, or advanced communication with a LIMS system, the Nimbus can meet the requirements. GLP printouts are available with time, date and other essential information.
Innovative design creates a smaller footprint, so the Nimbus occupies minimal space on the lab bench. Solid metal construction throughout the balance durably withstands chemicals and rigorous daily use. Optimized configuration of the Nimbus’s internal weighing system results in enhanced performance. Fabricated from a single block, the weighing sensor contains fewer parts than a traditional force motor balance, improving efficiency.
405,000.00INR / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment NBL 84 Analytical Balance, 80g x 0.1mg Backlit Lcd, 115V WW-11120-98 Mfr # NBL 84E-115V
COMPARE
External 80 0.0001
Call For Price
Adam Equipment NBL 84 Analytical Balance, 80g x 0.1mg Backlit Lcd, 230V WW-11120-99 Mfr # NBL 84E-230V
COMPARE
External 80 0.0001
Call For Price
Adam Equipment NBL-100 Analytical Balance, 100G x 0.1mg Backlit Lcd, 115V WW-11121-01 Mfr # NBL 104-115V
COMPARE
External 100 0.0001
Call For Price
Adam Equipment NBL-100 Analytical Balance, 100G x 0.1mg Backlit Lcd, 230V WW-11121-02 Mfr # NBL 104-230V
COMPARE
External 100 0.0001
Call For Price
Adam Equipment NBL 164 Analytical Balance, 160g x 0.1mg Backlit Lcd, 115V WW-11121-03 Mfr # NBL 164E-115V
COMPARE
External 160 0.0001
Call For Price
Adam Equipment NBL 164 Analytical Balance, 160g x 0.1mg Backlit Lcd, 230V WW-11121-04 Mfr # NBL 164E-230V
COMPARE
External 160 0.0001
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Adam Equipment NBL 124i Analytical Balance, 120 g x 0.1 mg with Internal Cal, 115V WW-11122-42 Mfr # NBL 124I 115V
COMPARE
Internal 120 0.0001
Call For Price
Adam Equipment NBL 124i Analytical Balance, 120 g x 0.1 mg with Internal Cal, 230V WW-11122-43 Mfr # NBL 124I 230V
COMPARE
Internal 120 0.0001
Call For Price
Adam Equipment NBL 164i Analytical Balance, 160 g x 0.1 mg , 115V WW-11122-50 Mfr # NBL 164I 115V
COMPARE
Internal 160 0.0001
Call For Price
Adam Equipment NBL 164i Analytical Balance, 160 g x 0.1 mg , 230V WW-11122-51 Mfr # NBL 164I 230V
COMPARE
Internal 160 0.0001
₹405,000.00INR / Each
Adam Equipment NBL 214i Analytical Balance, 210 g x 0.1 mg with Internal Cal, 115V WW-11122-54 Mfr # NBL 214I 115V
COMPARE
Internal 210 0.0001
Call For Price
Adam Equipment Nimbus NBL 214i Precision Toploading Balance, 210 g x 0.1 mg with Internal Cal, 230 V WW-11122-55 Mfr # NBL 214I 230V
COMPARE
Internal 210 0.0001
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Adam Equipment NBL 254i Analytical Balance, 250 g x 0.1 mg with Internal Cal, 115V WW-11122-62 Mfr # NBL 254I 115V
COMPARE
Internal 250 0.0001
Call For Price
Adam Equipment NBL 254i Analytical Balance, 250 g x 0.1 mg with Internal Cal, 230V WW-11122-63 Mfr # NBL 254I 230V
COMPARE
Internal 250 0.0001
Call For Price
Adam Equipment NBL 124E Analytical Balance, 120g x 0.1mg Backlit Lcd, 115V WW-11124-08 Mfr # NBL 124E - 115V
COMPARE
External 120 0.0001
Call For Price
Adam Equipment NBL 124E Analytical Balance, 120g x 0.1mg Backlit Lcd, 220V WW-11124-09 Mfr # NBL 124E - 220V
COMPARE
External 120 0.0001
Call For Price
Showing 1 - 20 of 26