This product is now discontinued, View Related Products
Adam Equipment Eclipse Toploading Balances
Simplifies the most complex weighing operations with intuitive operation
  • All-metal housing ensures rugged, durable protection and stability
  • GLP-compliant printouts via RS-232 or USB connection
  • Capacity tracker warns of approaching overloads
  • Weighing applications include parts counting, check weighing, check counting, net/total and density calculation, percent weighing and dynamic/animal weighing
  • Large multi-language backlit display with soft-touch keypad and illuminated keys
  • Below-balance weighing hook
Every Eclipse is outfitted with an elegant capacitive touch keypad containing colour-coded keys, which enable uncomplicated navigation of the functions and features. Operating ith a feather-light touch, the keys respond readily, even while wearing latex-gloves. The keypad’s smooth surface allows fingerprints to be wiped off easily. Illuminated keys guide users through tasks, highlighting available options.
Featuring one of the largest LCD readouts in the industry, the Eclipse displays sizeable white digits on a deep-blue background, ensuring ultimate visibility. A second line shows text prompts, instructions and any other relevant information. Clear, discernable symbols and multilingual
text make operation easy, while prompts simplify even the most complex activities.
The Eclipse achieves connectivity with precision and speed, whether it’s basic data printing or advanced communication with a Laboratory Information Management System. Printouts with
time, date, and other information are provided to comply with GLP requirements. USB and RS-232 interfaces optimise connections, while a third interface allows use of an optional remote display.
407,000.00 - ₹459,000.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment EBL 223e-115V Eclipse Precision Toploader Balance, 220G with 1mg Readability, Backlit Lcd, 115V, Graphic Lcdin WW-11121-36 Mfr # EBL 223E-115V
COMPARE
220 115 0.001
Call For Price
Adam Equipment EBL 223e-230V Eclipse Precision Toploader Balance, 220G with 1mg Readability, Backlit Lcd, 230V WW-11121-37 Mfr # EBL 223E-230V
COMPARE
220 230 0.001
Call For Price
Adam Equipment EBL 323e-115V Eclipse Precision Toploader Balance, 320G with 1mg Readability, Backlit Lcd, 115V WW-11121-38 Mfr # EBL 323E-115V
COMPARE
320 115 0.001
Call For Price
Adam Equipment EBL 323e-230V Eclipse Precision Toploader Balance, 320G with 1mg Readability, Backlit Lcd, 230V WW-11121-39 Mfr # EBL 323E-230V
COMPARE
320 230 0.001
Call For Price
Adam Equipment EBL 423e-115V Eclipse Precision Toploader Balance, 420G with 1mg Readability, Backlit Lcd, 115V WW-11121-40 Mfr # EBL 423E-115V
COMPARE
420 115 0.001
Call For Price
Adam Equipment EBL 423e-230V Eclipse Precision Toploader Balance, 420G with 1mg Readability, Backlit Lcd, 230V WW-11121-41 Mfr # EBL 423E-230V
COMPARE
420 230 0.001
Call For Price
Adam Equipment EBL 623e-115V Eclipse Precision Toploader Balance, 620G with 1mg Readability, Backlit Lcd, 115V WW-11121-42 Mfr # EBL 623E-115V
COMPARE
620 115 0.001
Call For Price
Adam Equipment EBL 623e-230V Eclipse Precision Toploader Balance, 620G with 1mg Readability, Backlit Lcd, 230V WW-11121-43 Mfr # EBL 623E-230V
COMPARE
620 230 0.001
Call For Price
Adam Equipment EBL 823e-115V Eclipse Precision Toploader Balance, 820G with 1mg Readability, Backlit Lcd, 115V WW-11121-44 Mfr # EBL 823E 115V
COMPARE
820 115 0.001
Call For Price
Adam Equipment EBL 823e-230V Eclipse Precision Toploader Balance, 820G with 1mg Readability, Backlit Lcd, 230V WW-11121-45 Mfr # EBL 823E 230V
COMPARE
820 230 0.001
Call For Price
Adam Equipment EBL 1023e-115V Eclipse Precision Toploader Balance, 1020G with 1mg Readability, Backlit Lcd, 115V WW-11121-46 Mfr # EBL 1023E-115V
COMPARE
1020 115 0.001
Call For Price
Adam Equipment EBL 1023e-230V Eclipse Precision Toploader Balance, 1020G with 1mg Readability, Backlit Lcd, 230V WW-11121-47 Mfr # EBL 1023E-230V
COMPARE
1020 230 0.001
Call For Price
Adam Equipment EBL 1623e-115V Eclipse Precision Toploader Balance, 1620G with 1mg Readability, Backlit Lcd, 115V WW-11121-48 Mfr # EBL 1623E-115V
COMPARE
1620 115 0.001
Call For Price
Adam Equipment EBL 1623e-230V Eclipse Precision Toploader Balance, 1620G with 1mg Readability, Backlit Lcd, 230V WW-11121-49 Mfr # EBL 1623E-230V
COMPARE
1620 230 0.001
Call For Price
Adam Equipment EBL 1602e-115V Eclipse Precision Toploader Balance, 1600G with 10mg Readability, Backlit Lcd, 115V WW-11121-50 Mfr # EBL 1602E-115V
COMPARE
1600 115 0.01
Call For Price
Adam Equipment EBL 1602e-230V Eclipse Precision Toploader Balance, 1600G with 10mg Readability, Backlit Lcd, 230V WW-11121-51 Mfr # EBL 1602E-230V
COMPARE
1600 230 0.01
Call For Price
Adam Equipment EBL 2602e-115V Eclipse Precision Toploader Balance, 2600G with 10mg Readability, Backlit Lcd, 115V WW-11121-52 Mfr # EBL 2602E-115V
COMPARE
2600 115 0.01
Call For Price
Adam Equipment EBL 2602e-230V Eclipse Precision Toploader Balance, 2600G with 10mg Readability, Backlit Lcd, 230V WW-11121-53 Mfr # EBL 2602E-230V
COMPARE
2600 230 0.01
Call For Price
Adam Equipment EBL 3602e-115V Eclipse Precision Toploader Balance, 3600G Capacity, with 10mg Readability, Backlit Lcd, 115V WW-11121-54 Mfr # EBL 3602E-115V
COMPARE
3600 115 0.01
Call For Price
Adam Equipment EBL 3602e-230V Eclipse Precision Toploader Balance, 3600G Capacity, with 10mg Readability, Backlit Lcd, 230V WW-11121-55 Mfr # EBL 3602E-230V
COMPARE
3600 230 0.01
Call For Price
Showing 1 - 20 of 50