This product is now discontinued, View Related Products
Adam Equipment Eclipse Analytical Balances
Simplifies the most complex weighing operations with intuitive operation
  • All-metal housing ensures rugged, durable protection and stability
  • GLP-compliant printouts via RS-232 or USB connection
  • Capacity tracker warns of approaching overloads
  • Weighing applications include parts counting, check weighing, check counting, net/total and density calculation, percent weighing and dynamic/animal weighing
  • Large multi-language backlit display with soft-touch keypad and illuminated keys
  • Weighing chamber can be easily disassembled for cleaning
  • Built-in memory stores date and time data
  • Below-balance weighing hook
Every Eclipse is outfitted with an elegant capacitive touch keypad containing colour-coded keys, which enable uncomplicated navigation of the functions and features. Operating ith a feather-light touch, the keys respond readily, even while wearing latex-gloves. The keypad’s smooth surface allows fingerprints to be wiped off easily. Illuminated keys guide users through tasks, highlighting available options.
Featuring one of the largest LCD readouts in the industry, the Eclipse displays sizeable white digits on a deep-blue background, ensuring ultimate visibility. A second line shows text prompts, instructions and any other relevant information. Clear, discernable symbols and multilingual
text make operation easy, while prompts simplify even the most complex activities.
The Eclipse achieves connectivity with precision and speed, whether it’s basic data printing or advanced communication with a Laboratory Information Management System. Printouts with
time, date, and other information are provided to comply with GLP requirements. USB and RS-232 interfaces optimise connections, while a third interface allows use of an optional remote display.
28 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Adam Equipment EBL 104e-115V Eclipse Analytical Balance, 100G with 0.1mg Readability, Backlit Lcd, 115V WW-11121-30 Mfr # EBL 104E-115V
COMPARE
External 100 0.0001
Call For Price
Adam Equipment EBL 104e-230V Eclipse Analytical Balance, 100G with 0.1mg Readability, Backlit Lcd, 230V WW-11121-31 Mfr # EBL 104E-230V
COMPARE
External 100 0.0001
Call For Price
Adam Equipment EBL 164e-115V Eclipse Analytical Balance, 160G with 0.1mg Readability, Backlit Lcd, 115V WW-11121-32 Mfr # EBL 164E-115V
COMPARE
External 160 0.0001
Call For Price
Adam Equipment EBL 164e-230V Eclipse Analytical Balance, 160G with 0.1mg Readability, Backlit Lcd, 230V WW-11121-33 Mfr # EBL 164E-230V
COMPARE
External 160 0.0001
Call For Price
Adam Equipment EBL 214e-115V Eclipse Analytical Balance, 210G with 0.1mg Readability, Backlit Lcd, 115V WW-11121-34 Mfr # EBL 214E-115V
COMPARE
External 210 0.0001
Call For Price
Adam Equipment EBL 214e-230V Eclipse Analytical Balance, 210G with 0.1mg Readability, Backlit Lcd, 230V WW-11121-35 Mfr # EBL 214E-230V
COMPARE
External 210 0.0001
Call For Price
Adam Equipment EBL 104i-115V Eclipse Analytical Balance, 100G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 115V WW-11121-74 Mfr # EBL 104I-115V
COMPARE
Internal 100 0.0001
Call For Price
Adam Equipment EBL 104i-230V Eclipse Analytical Balance, 100G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 230V WW-11121-75 Mfr # EBL 104I-230V
COMPARE
Internal 100 0.0001
Call For Price
Adam Equipment EBL 164i-115V Eclipse Analytical Balance, 160G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 115V WW-11121-76 Mfr # EBL 164I-115V
COMPARE
Internal 160 0.0001
Call For Price
Adam Equipment EBL 164i-230V Eclipse Analytical Balance, 160G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 230V WW-11121-77 Mfr # EBL 164I-230V
COMPARE
Internal 160 0.0001
Call For Price
Adam Equipment EBL 214i-115V Eclipse Analytical Balance, 210G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 115V WW-11121-78 Mfr # EBL 214I-115V
COMPARE
Internal 210 0.0001
Call For Price
Adam Equipment EBL 214i-230V Eclipse Analytical Balance, 210G with 0.1mg Readability, with Internal Cal, Backlit Lcd, 230V WW-11121-79 Mfr # EBL 214I-230V
COMPARE
Internal 210 0.0001
Call For Price
Adam Equipment EBL 254E Analytical Balance, 250 g x 0.1 mg, 115 V WW-11122-17 Mfr # EBL 254E 115V
COMPARE
Call For Price
Adam Equipment EBL 254E Analytical Balance, 250 g x 0.1 mg, 230 V WW-11122-19 Mfr # EBL 254E 230V
COMPARE
Call For Price
Adam Equipment EBL 254I Analytical Balance, 250 g x 0.1 mg, 115 V WW-11122-21 Mfr # EBL 254I 115V
COMPARE
Call For Price
Adam Equipment EBL 254I Analytical Balance, 250 g x 0.1 mg, 230 V WW-11122-23 Mfr # EBL 254I 230V
COMPARE
Call For Price
Adam Equipment EBL 314E Analytical Balance, 410 g x 0.1 mg, 115 V WW-11122-25 Mfr # EBL 314E 115V
COMPARE
Call For Price
Adam Equipment EBL 314E Analytical Balance, 410 g x 0.1 mg, 230 V WW-11122-27 Mfr # EBL 314E 230V
COMPARE
Call For Price
Adam Equipment EBL 314I Analytical Balance, 410 g x 0.1 mg, 115 V WW-11122-28 Mfr # EBL 314I 115V
COMPARE
Call For Price
Adam Equipment EBL 314I Analytical Balance, 410 g x 0.1 mg, 230 V WW-11122-29 Mfr # EBL 314I 230V
COMPARE
Call For Price
Showing 1 - 20 of 28