This product is now discontinued, View Related Products
Adam Equipment Astro ASC Stainless Steel Food Portioning Scales
Compact, sleek stainless steel scale is perfect for use in food industry
  • Solid, one-piece construction and a sealed keypad allow for quick wipe down
  • Large digits on the LCD change from green to red when the scale is overloaded
  • Color-coded keys provide quick access to the weighing units (g, oz, and lb)
  • Large, durable 304 stainless steel pan
  • Overload protection helps prevent damage
  • Feature low-battery indicator and auto shutoff
Astro portioning scales are designed to be durable enough to withstand the demands of a variety of food environments – from small café kitchens to large restaurant chains, from mid-sized catering companies to internationally known fast-food franchises. Readings on the display change from green to red to alert the user if the scale is overloaded. Astro's stainless steel housing is ideal for commercial kitchen settings, and allows efficient clean up with a simple wipe down after us.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment Astro ASC 2001 Stainless Steel Food Scale, 2000g x 0.1g 115V WW-11119-13 Mfr # ASC 2001-115V
COMPARE
2000 115 0.1
Call For Price
Adam Equipment Astro ASC 4001 Stainless Steel Food Scale, 4000g x 0.5g 115V WW-11119-14 Mfr # ASC 4001-115V
COMPARE
4000 115 0.5
Call For Price
Adam Equipment Astro ASC 8000 Stainless Steel Food Scale, 8000g x 1g 115V WW-11119-16 Mfr # ASC 8000-115V
COMPARE
8000 115 1
Call For Price
Adam Equipment Astro ASC 2001 Stainless Steel Food Scale, 2000g x 0.1g 220V WW-11119-17 Mfr # ASC 2001-220V
COMPARE
2000 220 0.1
Call For Price
Adam Equipment Astro ASC 4001 Stainless Steel Food Scale, 4000g x 0.5g 220V WW-11119-18 Mfr # ASC 4001-220V
COMPARE
4000 220 0.5
Call For Price
Adam Equipment Astro ASC 8000 Stainless Steel Food Scale, 8000g x 1g 220V WW-11119-19 Mfr # ASC 8000-220V
COMPARE
8000 220 1