Advantec Aborbent Cellulose Pads for Petri Dishes
Ideal for use in culturing microorganisms
  • Tested for absorption of 1.8 to 2.2 mL of Culture media
  • pH neutral
  • Individually packaged to assure sterility
Absorbent pads are white cellulose discs that fit into standard petri dishes. Pre-sterilized and tested for absorption of 1.8 to 2.2 mL of culture media, these pads are ideal for culturing using broth media in petri dishes.
6,020.00INR / Box of 100
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Max Absorption
Availability Pricing
Advantec Aborbent Pads for Petri Dishes, Sterile, Cellulose, 47 mm; 100/Bx WW-14005-22 Mfr # B200G047A
COMPARE
47 Cellulose 1.8 to 2.2 mL
₹6,020.00INR / Box of 100